accessibility menu, dialog, popup

UserName

CAS RN 925-78-0

IUPAC Name: nonan-3-one
Synonyms: 3-nonanone
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: IYTXKIXETAELAV-UHFFFAOYSA-N
SMILES: CCCCCCC(=O)CC
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)