missing translation for 'onlineSavingsMsg'
Learn More

3-Nonanol, 98%

CAS: 624-51-1 | C9H20O | 144.258 g/mol

Supplier:  Thermo Scientific Chemicals A1912018

 View more versions of this product

Catalog No. AAA1912018


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Nonanol
0.824
79°C (174°F)
1.431
50 g
3-nonanol, hexylethylcarbinol, ethylhexylcarbinol, 3-nonyl alcohol, nonan-7-ol, ethyl hexyl carbinol, acmc-1b2h7
CCCCCCC(CC)O
144.258
144.26
624-51-1
192°C to 194°C
C9H20O
MFCD00014411
1719453
GYSCXPVAKHVAAY-UHFFFAOYSA-N
nonan-3-ol
12216
98%

Chemical Identifiers

624-51-1
144.258
GYSCXPVAKHVAAY-UHFFFAOYSA-N
12216
CCCCCCC(CC)O
C9H20O
MFCD00014411
3-nonanol, hexylethylcarbinol, ethylhexylcarbinol, 3-nonyl alcohol, nonan-7-ol, ethyl hexyl carbinol, acmc-1b2h7
nonan-3-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 210-850-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only