missing translation for 'onlineSavingsMsg'
Learn More

3-Nonanol, 98%

Catalog No. p-7024449
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1912018 50 g
AAA1912030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1912018 Supplier Thermo Scientific Chemicals Supplier No. A1912018

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 624-51-1 | C9H20O | 144.258 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 624-51-1
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.258
MDL Number MFCD00014411
InChI Key GYSCXPVAKHVAAY-UHFFFAOYSA-N
Synonym 3-nonanol, hexylethylcarbinol, ethylhexylcarbinol, 3-nonyl alcohol, nonan-7-ol, ethyl hexyl carbinol, acmc-1b2h7
PubChem CID 12216
IUPAC Name nonan-3-ol
SMILES CCCCCCC(CC)O

Specifications

Density 0.824
Boiling Point 192°C to 194°C
Flash Point 79°C (174°F)
Refractive Index 1.431
Quantity 50 g
Beilstein 1719453
Formula Weight 144.26
Percent Purity 98%
Chemical Name or Material 3-Nonanol
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 210-850-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only