missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrotoluene 99.0+%, TCI America™

Catalog No. N0274500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0274500G 500 g
N027425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0274500G Supplier TCI America Supplier No. N0274500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-08-1
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007265
InChI Key QZYHIOPPLUPUJF-UHFFFAOYSA-N
Synonym 3-nitrotoluene, m-nitrotoluene, 3-nitrotoluol, benzene, 1-methyl-3-nitro, m-methylnitrobenzene, toluene, m-nitro, meta-nitrotoluol, 3-methylnitrobenzene, m-nitrotoluol, nitrotoluene, m
PubChem CID 7422
ChEBI CHEBI:39931
IUPAC Name 1-methyl-3-nitrobenzene
SMILES CC1=CC=CC(=C1)[N+]([O-])=O

Specifications

Color Yellow
Boiling Point 232°C
Quantity 500 g
UN Number 1664
Formula Weight 137.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Nitrotoluene
EINECSNumber (3)-0437
RTECSNumber XT2975000
TSCA Yes