missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrotoluene, 99%

Catalog No. AC129045000
Change view
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC129045000 500 mL Glass bottle
AC129040050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC129045000 Supplier Thermo Scientific Chemicals Supplier No. 129045000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-08-1 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-08-1
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007265
InChI Key QZYHIOPPLUPUJF-UHFFFAOYSA-N
Synonym 3-nitrotoluene, m-nitrotoluene, 3-nitrotoluol, benzene, 1-methyl-3-nitro, m-methylnitrobenzene, toluene, m-nitro, meta-nitrotoluol, 3-methylnitrobenzene, m-nitrotoluol, nitrotoluene, m
PubChem CID 7422
ChEBI CHEBI:39931
IUPAC Name 1-methyl-3-nitrobenzene
SMILES CC1=CC=CC(=C1)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 15°C to 16°C
Color Brown-Yellow to Yellow
Density 1.1500g/mL
Boiling Point 230°C to 231°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5450 to 1.547
Linear Formula CH3C6H4NO2
Quantity 500 mL
Beilstein 05, 321
Specific Gravity 1.15
Merck Index 15, 6737
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and benzene,miscible with ether
Formula Weight 137.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Nitrotoluene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-728-6
RTECSNumber XT2975000
TSCA TSCA

RUO – Research Use Only