missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrothiobenzamide, 97%

Catalog No. AAH5215206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5215206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5215206 Supplier Thermo Scientific Chemicals Supplier No. H5215206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70102-34-0 | C7H6N2O2S | 182.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70102-34-0
Molecular Formula C7H6N2O2S
Molecular Weight (g/mol) 182.197
MDL Number MFCD09757586
InChI Key HDQCHDWHHGEXQE-UHFFFAOYSA-N
Synonym 3-nitrobenzene-1-carbothioamide, 3-nitrothiobenzamide, 3-nitrobenzothioamide, 3-nitro-thiobenzamide, 3-nitrobenzenethioamide, 3-nitro-thio-benzamide, benzenecarbothioamide, 3-nitro
PubChem CID 12668182
IUPAC Name 3-nitrobenzenecarbothioamide
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=S)N

Specifications

Quantity 5 g
Formula Weight 182.2
Percent Purity 97%
Chemical Name or Material 3-Nitrothiobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only