missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrosalicylic Acid 98.0+%, TCI America™

Catalog No. p-7133704
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02515G 5 g
N025125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02515G Supplier TCI America Supplier No. N02515G
Only null left

Chemical Identifiers

CAS 85-38-1
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.119
MDL Number MFCD00024240
InChI Key WWWFHFGUOIQNJC-UHFFFAOYSA-N
Synonym 3-nitrosalicylic acid, 2-hydroxy-3-nitrobenzoic acid, benzoic acid, 2-hydroxy-3-nitro, salicylic acid, 3-nitro, 2-hydroxy-3-nitro-benzoic acid, 3-nitro-salicylsaure, 3-nitrosalicyclic acid, wln: wnr bq cvq, acmc-209q5q
PubChem CID 6807
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])O)C(=O)O

Specifications

Melting Point 149°C
Quantity 5 g
Formula Weight 183.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrosalicylic Acid
RTECSNumber VO5300000
TSCA No