missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrosalicylic acid, 98%

Catalog No. p-6827180
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399820010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399820010 Supplier Thermo Scientific Chemicals Supplier No. 399820010

CAS: 85-38-1 | C7H5NO5 | 183.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85-38-1
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
InChI Key WWWFHFGUOIQNJC-UHFFFAOYSA-N
Synonym 3-nitrosalicylic acid, 2-hydroxy-3-nitrobenzoic acid, benzoic acid, 2-hydroxy-3-nitro, salicylic acid, 3-nitro, 2-hydroxy-3-nitro-benzoic acid, 3-nitro-salicylsaure, 3-nitrosalicyclic acid, wln: wnr bq cvq, acmc-209q5q
PubChem CID 6807
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])O)C(=O)O

Specifications

Melting Point 142°C to 147°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula O2NC6H3(OH)COOH
Quantity 1 g
Merck Index 15,6717
Formula Weight 183.12
Percent Purity 98%
Chemical Name or Material 3-Nitrosalicylic acid
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only