missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalonitrile 98.0+%, TCI America™

Catalog No. N05665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05665G 5 g
N056625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05665G Supplier TCI America Supplier No. N05665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51762-67-5
Molecular Formula C8H3N3O2
Molecular Weight (g/mol) 173.13
MDL Number MFCD00191558
InChI Key UZJZIZFCQFZDHP-UHFFFAOYSA-N
Synonym 3-nitrophthalonitrile, 2,3-dicyanonitrobenzene, 1,2-benzenedicarbonitrile, 3-nitro, 3-nitrophthalodinitrile, 1,2-dicyano-3-nitrobenzene, 3-nitro-1,2-benzenedicarbonitrile, nitrophthalonitrile, 3-nitro-phthalonitrile, 3-nitrophthalonitrile?, acmc-209kvo
PubChem CID 162652
IUPAC Name 3-nitrobenzene-1,2-dicarbonitrile
SMILES [O-][N+](=O)C1=CC=CC(C#N)=C1C#N

Specifications

Melting Point 163°C
Quantity 5 g
Formula Weight 173.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrophthalonitrile
RTECSNumber CZ1953200
TSCA No