missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalic Anhydride 97.0+%, TCI America™

Catalog No. p-7133698
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N024525G 25 g
N0245250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N024525G Supplier TCI America Supplier No. N024525G
Only null left

Chemical Identifiers

CAS 641-70-3
Molecular Formula C8H3NO5
Molecular Weight (g/mol) 193.11
MDL Number MFCD00005921
InChI Key ROFZMKDROVBLNY-UHFFFAOYSA-N
Synonym 3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride
PubChem CID 21631
IUPAC Name 4-nitro-1,3-dihydro-2-benzofuran-1,3-dione
SMILES [O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O

Specifications

Melting Point 162°C
Quantity 25 g
Formula Weight 193.11
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrophthalic Anhydride