missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalic anhydride, 97%

Catalog No. AAA1585722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1585722 100 g
AAA1585714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1585722 Supplier Thermo Scientific Chemicals Supplier No. A1585722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 641-70-3 | C8H3NO5 | 193.11 g/mol

4-Nitrophthalic Anhydride is an intermediate for the synthesis of the benzimidazole PARP inhibitor I, ABT-472. It reacts with aminoquinazolinones to yield phthalimidoquinazolinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophthalic Anhydride is an intermediate for the synthesis of the benzimidazole PARP inhibitor I, ABT-472. It reacts with aminoquinazolinones to yield phthalimidoquinazolinones.

Solubility
May decompose in water.

Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 641-70-3
Molecular Formula C8H3NO5
Molecular Weight (g/mol) 193.11
MDL Number MFCD00005921
InChI Key ROFZMKDROVBLNY-UHFFFAOYSA-N
Synonym 3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride
PubChem CID 21631
SMILES [O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O

Specifications

Melting Point 161°C to 166°C
Quantity 100 g
Beilstein 179963
Sensitivity Moisture sensitive
Solubility Information May decompose in water.
IUPAC Name 4-nitro-2-benzofuran-1,3-dione
Formula Weight 193.11
Percent Purity 97%
Chemical Name or Material 3-Nitrophthalic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-373-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only