missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenylsulfur Pentafluoride 96.0+%, TCI America™

Catalog No. N07421G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N07421G Supplier TCI America Supplier No. N07421G

Chemical Identifiers

CAS 2613-26-5
Molecular Formula C6H4F5NO2S
Molecular Weight (g/mol) 249.155
MDL Number MFCD00221621
InChI Key FSTNQYCPXJMFMT-UHFFFAOYSA-N
Synonym 3-nitrophenylsulfur pentafluoride, 3-nitrophenylsulphur pentafluoride, 1-nitro-3-pentafluorosulfanyl benzene, 1-nitro-3-pentafluoro-??-sulfanyl benzene, acmc-209goj, pentafluoro-3-nitrophenyl, 3-pentafluorothio nitrobenzene, 3-nitropentafluorosulfanylbenzene, 3-pentafluorosulphanylnitrobenzene, 3-pentafluorosulfur-nitrobenzene
PubChem CID 2775738
IUPAC Name pentafluoro-(3-nitrophenyl)-$l^{6}-sulfane
SMILES C1=CC(=CC(=C1)S(F)(F)(F)(F)F)[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 83°C
Quantity 1 g
UN Number 2810
Formula Weight 249.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Nitrophenylsulfur Pentafluoride