missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenylhydrazine Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America N023225G

 View more versions of this product

Catalog No. N023225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Nitrophenylhydrazine Hydrochloride
C6H8ClN3O2
25 g
3-nitrophenylhydrazine hydrochloride, 3-nitrophenyl hydrazine hydrochloride, 3-nitrophenylhydrazinium 1+ chloride, hydrazine, 3-nitrophenyl-, monohydrochloride, m-nitrophenylhydrazine hydrochloride, pubchem20000, acmc-1b1e3, ksc491e2l, m-nitrophenylhydrazine hcl
C1=CC(=CC(=C1)[N+](=O)[O-])NN.Cl
189.599
189.60
Crystalline Powder
636-95-3
MFCD00012939
1325
BKOYKMLGFFASBG-UHFFFAOYSA-N
(3-nitrophenyl)hydrazine;hydrochloride
5464285
≥98.0% (T)

Chemical Identifiers

636-95-3
189.599
BKOYKMLGFFASBG-UHFFFAOYSA-N
5464285
C1=CC(=CC(=C1)[N+](=O)[O-])NN.Cl
C6H8ClN3O2
MFCD00012939
3-nitrophenylhydrazine hydrochloride, 3-nitrophenyl hydrazine hydrochloride, 3-nitrophenylhydrazinium 1+ chloride, hydrazine, 3-nitrophenyl-, monohydrochloride, m-nitrophenylhydrazine hydrochloride, pubchem20000, acmc-1b1e3, ksc491e2l, m-nitrophenylhydrazine hcl
(3-nitrophenyl)hydrazine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0477

TSCA : No

SDS
Documents

Documents

Promotions

Promotions