missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. N02325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02325G 5 g
N023225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02325G Supplier TCI America Supplier No. N02325G
Only null left

Chemical Identifiers

CAS 636-95-3
Molecular Formula C6H8ClN3O2
Molecular Weight (g/mol) 189.599
MDL Number MFCD00012939
InChI Key BKOYKMLGFFASBG-UHFFFAOYSA-N
Synonym 3-nitrophenylhydrazine hydrochloride, 3-nitrophenyl hydrazine hydrochloride, 3-nitrophenylhydrazinium 1+ chloride, hydrazine, 3-nitrophenyl-, monohydrochloride, m-nitrophenylhydrazine hydrochloride, pubchem20000, acmc-1b1e3, ksc491e2l, m-nitrophenylhydrazine hcl
PubChem CID 5464285
IUPAC Name (3-nitrophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])NN.Cl

Specifications

Quantity 5 g
UN Number 1325
Formula Weight 189.60
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrophenylhydrazine Hydrochloride
EINECSNumber (3)-0477
TSCA No