missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrochalcone 98.0+%, TCI America™
$137.94 - $456.32
Chemical Identifiers
| CAS | 614-48-2 |
|---|---|
| Molecular Formula | C15H11NO3 |
| Molecular Weight (g/mol) | 253.257 |
| MDL Number | MFCD00024571 |
| InChI Key | SMFBODMWKWBFOK-MDZDMXLPSA-N |
| Synonym | 3-nitrochalcone, chalcone, 3-nitro, m-nitrobenzylidene acetophenone, 2-propen-1-one, 3-3-nitrophenyl-1-phenyl, 3-nitrobenzylideneacetophenone, ccris 1670, 2e-3-3-nitrophenyl-1-phenylprop-2-en-1-one, e-3-3-nitrophenyl-1-phenyl-2-propen-1-one, e-3-3-nitrophenyl-1-phenylprop-2-en-1-one, e-3-3-nitrophenyl-1-phenyl-prop-2-en-1-one |
| PubChem CID | 5369664 |
| IUPAC Name | (E)-3-(3-nitrophenyl)-1-phenylprop-2-en-1-one |
| SMILES | C1=CC=C(C=C1)C(=O)C=CC2=CC(=CC=C2)[N+](=O)[O-] |
Chemical Identifiers
| 614-48-2 | |
| 253.257 | |
| SMFBODMWKWBFOK-MDZDMXLPSA-N | |
| 5369664 | |
| C1=CC=C(C=C1)C(=O)C=CC2=CC(=CC=C2)[N+](=O)[O-] |
| C15H11NO3 | |
| MFCD00024571 | |
| 3-nitrochalcone, chalcone, 3-nitro, m-nitrobenzylidene acetophenone, 2-propen-1-one, 3-3-nitrophenyl-1-phenyl, 3-nitrobenzylideneacetophenone, ccris 1670, 2e-3-3-nitrophenyl-1-phenylprop-2-en-1-one, e-3-3-nitrophenyl-1-phenyl-2-propen-1-one, e-3-3-nitrophenyl-1-phenylprop-2-en-1-one, e-3-3-nitrophenyl-1-phenyl-prop-2-en-1-one | |
| (E)-3-(3-nitrophenyl)-1-phenylprop-2-en-1-one |
Specifications
| 614-48-2 | |
| Yellow | |
| MFCD00024571 | |
| 3-nitrochalcone, chalcone, 3-nitro, m-nitrobenzylidene acetophenone, 2-propen-1-one, 3-3-nitrophenyl-1-phenyl, 3-nitrobenzylideneacetophenone, ccris 1670, 2e-3-3-nitrophenyl-1-phenylprop-2-en-1-one, e-3-3-nitrophenyl-1-phenyl-2-propen-1-one, e-3-3-nitrophenyl-1-phenylprop-2-en-1-one, e-3-3-nitrophenyl-1-phenyl-prop-2-en-1-one | |
| C1=CC=C(C=C1)C(=O)C=CC2=CC(=CC=C2)[N+](=O)[O-] | |
| 253.257 | |
| 253.26 | |
| Crystalline Powder |
| 144°C | |
| C15H11NO3 | |
| 1 g | |
| SMFBODMWKWBFOK-MDZDMXLPSA-N | |
| (E)-3-(3-nitrophenyl)-1-phenylprop-2-en-1-one | |
| 5369664 | |
| ≥98.0% (GC) | |
| 3-Nitrochalcone |
Safety and Handling
RTECSNumber : FL7075490
TSCA : No