missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzylamine hydrochloride, 97%

CAS: 26177-43-5 | C7H8N2O2·HCl | 188.62 g/mol

$426.28 - $426.28

Chemical Identifiers

CAS 26177-43-5
Molecular Formula C7H8N2O2·HCl
Molecular Weight (g/mol) 188.62
MDL Number MFCD00012858
InChI Key DLZXLCHQWOZGSE-UHFFFAOYSA-N
Synonym 3-nitrobenzylamine hydrochloride, 3-nitrophenyl methanamine hydrochloride, 3-nitrobenzylamine hcl, 3-nitrobenzylammonium hydrochloride, benzenemethanamine, 3-nitro-, monohydrochloride, 1-3-nitrophenyl methanamine hydrochloride, pubchem7485, chembl13978, m-nitrobenzylamine hydrochloride, 3-nitrobenzyl amine hydrochloride
PubChem CID 2724030
IUPAC Name (3-nitrophenyl)methanamine;hydrochloride
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC184730050
SDS View Documents
Thermo Scientific Chemicals
184730050
5 g Glass bottle
Each for $426.28
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

26177-43-5
188.62
DLZXLCHQWOZGSE-UHFFFAOYSA-N
2724030
C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl
C7H8N2O2·HCl
MFCD00012858
3-nitrobenzylamine hydrochloride, 3-nitrophenyl methanamine hydrochloride, 3-nitrobenzylamine hcl, 3-nitrobenzylammonium hydrochloride, benzenemethanamine, 3-nitro-, monohydrochloride, 1-3-nitrophenyl methanamine hydrochloride, pubchem7485, chembl13978, m-nitrobenzylamine hydrochloride, 3-nitrobenzyl amine hydrochloride
(3-nitrophenyl)methanamine;hydrochloride

Specifications

26177-43-5
100.0
Tan to White
97%
C7H8N2O2·HCl
MFCD00012858
12, 1076
DLZXLCHQWOZGSE-UHFFFAOYSA-N
(3-nitrophenyl)methanamine;hydrochloride
2724030
97%
3-Nitrobenzylamine hydrochloride, 97%
96.0
226°C to 231°C
Authentic
Glass bottle
O2NC6H4CH2NH2·HCl
5 g
3-nitrobenzylamine hydrochloride, 3-nitrophenyl methanamine hydrochloride, 3-nitrobenzylamine hcl, 3-nitrobenzylammonium hydrochloride, benzenemethanamine, 3-nitro-, monohydrochloride, 1-3-nitrophenyl methanamine hydrochloride, pubchem7485, chembl13978, m-nitrobenzylamine hydrochloride, 3-nitrobenzyl amine hydrochloride
C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl
188.62
188.62
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 247-502-8

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only