missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrobenzyl Chloride 99.0+%, TCI America™
$85.75 - $262.83
Chemical Identifiers
| CAS | 619-23-8 |
|---|---|
| Molecular Formula | C7H6ClNO2 |
| Molecular Weight (g/mol) | 171.58 |
| MDL Number | MFCD00007272 |
| InChI Key | APGGSERFJKEWFG-UHFFFAOYSA-N |
| Synonym | 3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324 |
| PubChem CID | 12078 |
| IUPAC Name | 1-(chloromethyl)-3-nitrobenzene |
| SMILES | [O-][N+](=O)C1=CC=CC(CCl)=C1 |
Chemical Identifiers
| 619-23-8 | |
| 171.58 | |
| APGGSERFJKEWFG-UHFFFAOYSA-N | |
| 12078 | |
| [O-][N+](=O)C1=CC=CC(CCl)=C1 |
| C7H6ClNO2 | |
| MFCD00007272 | |
| 3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324 | |
| 1-(chloromethyl)-3-nitrobenzene |
Specifications
| 619-23-8 | |
| Yellow | |
| C7H6ClNO2 | |
| 5 g | |
| 3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324 | |
| [O-][N+](=O)C1=CC=CC(CCl)=C1 | |
| 171.58 | |
| 171.58 | |
| Crystalline Powder |
| 46°C | |
| 174°C | |
| MFCD00007272 | |
| 3261 | |
| APGGSERFJKEWFG-UHFFFAOYSA-N | |
| 1-(chloromethyl)-3-nitrobenzene | |
| 12078 | |
| ≥99.0% (GC) | |
| 3-Nitrobenzyl Chloride |
Safety and Handling
EINECSNumber : (3)-0449
RTECSNumber : XS9090000
TSCA : Yes