missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzyl Chloride 99.0+%, TCI America™

Catalog No. C07095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07095G 5 g
C070925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07095G Supplier TCI America Supplier No. C07095G

Chemical Identifiers

CAS 619-23-8
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007272
InChI Key APGGSERFJKEWFG-UHFFFAOYSA-N
Synonym 3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324
PubChem CID 12078
IUPAC Name 1-(chloromethyl)-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(CCl)=C1

Specifications

Melting Point 46°C
Color Yellow
Boiling Point 174°C
Quantity 5 g
UN Number 3261
Formula Weight 171.58
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrobenzyl Chloride
EINECSNumber (3)-0449
RTECSNumber XS9090000
TSCA Yes