missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzyl chloride, 98%

CAS: 619-23-8 | C7H6ClNO2 | 171.58 g/mol

$83.49 - $810.27

Chemical Identifiers

CAS 619-23-8
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007272
InChI Key APGGSERFJKEWFG-UHFFFAOYSA-N
Synonym 3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324
PubChem CID 12078
IUPAC Name 1-(chloromethyl)-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(CCl)=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1677509
View Documents
Thermo Scientific Chemicals
A1677509
10 g
Each for $83.49
Only null left
 
AAA1677518
View Documents
Thermo Scientific Chemicals
A1677518
50 g
Each for $248.90
Only null left
 
AAA1677530
View Documents
Thermo Scientific Chemicals
A1677530
250 g
Each for $810.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

619-23-8
171.58
APGGSERFJKEWFG-UHFFFAOYSA-N
12078
[O-][N+](=O)C1=CC=CC(CCl)=C1
C7H6ClNO2
MFCD00007272
3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324
1-(chloromethyl)-3-nitrobenzene

Specifications

619-23-8
150°C (3 mmHg)
C7H6ClNO2
10 g
742794
APGGSERFJKEWFG-UHFFFAOYSA-N
1-(chloromethyl)-3-nitrobenzene
12078
98%
43°C to 47°C
>110°C (230°F)
MFCD00007272
UN3261
3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324
[O-][N+](=O)C1=CC=CC(CCl)=C1
171.58
171.58
3-Nitrobenzyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 210-586-1

RTECSNumber : XS9090000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only