missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzyl chloride, 98%

Catalog No. AAA1677509
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1677509 10 g
AAA1677518 50 g
AAA1677530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1677509 Supplier Thermo Scientific Chemicals Supplier No. A1677509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 619-23-8 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-23-8
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007272
InChI Key APGGSERFJKEWFG-UHFFFAOYSA-N
Synonym 3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324
PubChem CID 12078
IUPAC Name 1-(chloromethyl)-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(CCl)=C1

Specifications

Melting Point 43°C to 47°C
Boiling Point 150°C (3 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN3261
Beilstein 742794
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 210-586-1
RTECSNumber XS9090000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only