missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzyl Alcohol 99.0+%, TCI America™

Catalog No. p-7135871
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03781G 1 g
S037810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03781G Supplier TCI America Supplier No. S03781G
Only null left

Chemical Identifiers

CAS 619-25-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007273
InChI Key CWNPOQFCIIFQDM-UHFFFAOYSA-N
Synonym 3-nitrobenzyl alcohol, 3-nitrophenyl methanol, m-nitrobenzyl alcohol, benzenemethanol, 3-nitro, benzyl alcohol, m-nitro, 3-nitrobenzenemethanol, unii-f829x990iv, ccris 7971, 3-nitrophenyl methan-1-ol, 3-nitrobenzylalcohol
PubChem CID 69267
IUPAC Name (3-nitrophenyl)methanol
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CO

Specifications

Quantity 1 g
Formula Weight 153.14
Percent Purity ≥99.0% (GC)
Physical Form Crystals
Chemical Name or Material 3-Nitrobenzyl Alcohol [Matrix for FABMS and liquid SIMS]
RTECSNumber DP0657000
TSCA Yes