Learn More
3-Nitrobenzoyl chloride, 98%
CAS: 121-90-4 | C7H4ClNO3 | 185.563 g/mol
Supplier: Thermo Scientific Chemicals A105790B
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Nitrobenzoyl chloride | |
| 31°C to 35°C | |
| 276°C to 278°C (decomposition) | |
| Pungent | |
| MFCD00007247 | |
| UN3261 | |
| Moisture sensitive | |
| NXTNASSYJUXJDV-UHFFFAOYSA-N | |
| 3-nitrobenzoyl chloride | |
| 8495 | |
| 98% |
| 121-90-4 | |
| 1.42 | |
| >110°C (230°F) | |
| C7H4ClNO3 | |
| 1000 g | |
| 777186 | |
| m-nitrobenzoyl chloride, benzoyl chloride, 3-nitro, benzoyl chloride, m-nitro, 3-nitro-benzoyl chloride, ccris 1186, m-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove, 3-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove czech, m-nitrobenzoylchlorid | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl | |
| 185.563 | |
| 185.57 |
Chemical Identifiers
| 121-90-4 | |
| 185.563 | |
| NXTNASSYJUXJDV-UHFFFAOYSA-N | |
| 8495 | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl |
| C7H4ClNO3 | |
| MFCD00007247 | |
| m-nitrobenzoyl chloride, benzoyl chloride, 3-nitro, benzoyl chloride, m-nitro, 3-nitro-benzoyl chloride, ccris 1186, m-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove, 3-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove czech, m-nitrobenzoylchlorid | |
| 3-nitrobenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H311-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 204-505-9
RTECSNumber : DM6646000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only