missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzotrifluoride 96.0+%, TCI America™

Catalog No. N028025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N028025G 25 g
N0280500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N028025G Supplier TCI America Supplier No. N028025G

Chemical Identifiers

CAS 98-46-4
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
MDL Number MFCD00007260
InChI Key WHNAMGUAXHGCHH-UHFFFAOYSA-N
Synonym 3-nitrobenzotrifluoride, 1-nitro-3-trifluoromethyl benzene, m-nitrobenzotrifluoride, 3-trifluoromethyl nitrobenzene, m-nitrotrifluorotoluene, benzene, 1-nitro-3-trifluoromethyl, 1,3-nitrobenzotrifluoride, m-nitrotrifluortoluol, usaf ma-5, 3-trifluoromethylnitrobenzene
PubChem CID 7386
IUPAC Name 1-nitro-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(F)(F)F

Specifications

Color Yellow
Boiling Point 203°C
Quantity 25 g
UN Number 2306
Formula Weight 191.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Nitrobenzotrifluoride
EINECSNumber (3)-0443
RTECSNumber XT3500000
TSCA Yes