Learn More
3-Nitrobenzotrifluoride, 97%
CAS: 98-46-4 | C7H4F3NO2 | 191.109 g/mol
$194.18 - $194.18
Chemical Identifiers
| CAS | 98-46-4 |
|---|---|
| Molecular Formula | C7H4F3NO2 |
| Molecular Weight (g/mol) | 191.109 |
| MDL Number | MFCD00007260 |
| InChI Key | WHNAMGUAXHGCHH-UHFFFAOYSA-N |
| Synonym | 3-nitrobenzotrifluoride, 1-nitro-3-trifluoromethyl benzene, m-nitrobenzotrifluoride, 3-trifluoromethyl nitrobenzene, m-nitrotrifluorotoluene, benzene, 1-nitro-3-trifluoromethyl, 1,3-nitrobenzotrifluoride, m-nitrotrifluortoluol, usaf ma-5, 3-trifluoromethylnitrobenzene |
| PubChem CID | 7386 |
| IUPAC Name | 1-nitro-3-(trifluoromethyl)benzene |
| SMILES | C1=CC(=CC(=C1)[N+](=O)[O-])C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0594822
|
Thermo Scientific Chemicals
L0594822 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 98-46-4 | |
| 191.109 | |
| WHNAMGUAXHGCHH-UHFFFAOYSA-N | |
| 7386 | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C(F)(F)F |
| C7H4F3NO2 | |
| MFCD00007260 | |
| 3-nitrobenzotrifluoride, 1-nitro-3-trifluoromethyl benzene, m-nitrobenzotrifluoride, 3-trifluoromethyl nitrobenzene, m-nitrotrifluorotoluene, benzene, 1-nitro-3-trifluoromethyl, 1,3-nitrobenzotrifluoride, m-nitrotrifluortoluol, usaf ma-5, 3-trifluoromethylnitrobenzene | |
| 1-nitro-3-(trifluoromethyl)benzene |
Specifications
| 98-46-4 | |
| 1.435 | |
| 87°C (188°F) | |
| C7H4F3NO2 | |
| MFCD00007260 | |
| UN2306 | |
| 3-nitrobenzotrifluoride, 1-nitro-3-trifluoromethyl benzene, m-nitrobenzotrifluoride, 3-trifluoromethyl nitrobenzene, m-nitrotrifluorotoluene, benzene, 1-nitro-3-trifluoromethyl, 1,3-nitrobenzotrifluoride, m-nitrotrifluortoluol, usaf ma-5, 3-trifluoromethylnitrobenzene | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C(F)(F)F | |
| 191.109 | |
| 191.11 | |
| 3-Nitrobenzotrifluoride |
| -5°C | |
| 201°C to 205°C | |
| Characteristic | |
| 1.472 | |
| 100 g | |
| 2051362 | |
| WHNAMGUAXHGCHH-UHFFFAOYSA-N | |
| 1-nitro-3-(trifluoromethyl)benzene | |
| 7386 | |
| 97% |
Safety and Handling
GHS H Statement
H301-H311-H330-H227-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312-H315-H319-H330-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROBENZOTRIFLUORIDES, LIQUID
EINECSNumber : 202-670-1
RTECSNumber : XT3500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only