missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzonitrile 99.0+%, TCI America™

Catalog No. N017025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N017025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N017025G Supplier TCI America Supplier No. N017025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 619-24-9
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.121
MDL Number MFCD00007194
InChI Key RUSAWEHOGCWOPG-UHFFFAOYSA-N
Synonym benzonitrile, 3-nitro, m-nitrobenzonitrile, 3-cyanonitrobenzene, m-cyanonitrobenzene, benzonitrile, m-nitro, 3-cyano-1-nitrobenzene, 3-nitro-benzonitrile, ccris 2327, 3-nitrobenzenecarbonitrile, 3-nitrobenzonitril
PubChem CID 12079
IUPAC Name 3-nitrobenzonitrile
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C#N

Specifications

Melting Point 115°C
Color Yellow
Boiling Point 165°C
Quantity 25 g
UN Number 3439
Formula Weight 148.12
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrobenzonitrile
EINECSNumber (3)-4141
RTECSNumber DI4900000
TSCA Yes