missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzonitrile, 98%

Catalog No. AC173520050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173520050 5 g Glass bottle
AC173520250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC173520050 Supplier Thermo Scientific Chemicals Supplier No. 173520050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 619-24-9 | C7H4N2O2 | 148.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-24-9
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.12
MDL Number MFCD00007194
InChI Key RUSAWEHOGCWOPG-UHFFFAOYSA-N
Synonym benzonitrile, 3-nitro, m-nitrobenzonitrile, 3-cyanonitrobenzene, m-cyanonitrobenzene, benzonitrile, m-nitro, 3-cyano-1-nitrobenzene, 3-nitro-benzonitrile, ccris 2327, 3-nitrobenzenecarbonitrile, 3-nitrobenzonitril
PubChem CID 12079
IUPAC Name 3-nitrobenzonitrile
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 115°C to 119°C
Color Yellow
Boiling Point 165°C (21.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula O2NC6H4CN
Quantity 5 g
Beilstein 09, 385
Solubility Information Solubility in water: insoluble. Other solubilities: freele soluble in ether and alcohol
Formula Weight 148.12
Percent Purity 98%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 3-Nitrobenzonitrile, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash w
EINECSNumber 210-587-7
RTECSNumber DI4900000
TSCA TSCA

RUO – Research Use Only