missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrobenzamide 98.0+%, TCI America™
$115.57 - $932.03
Chemical Identifiers
| CAS | 645-09-0 |
|---|---|
| Molecular Formula | C7H6N2O3 |
| Molecular Weight (g/mol) | 166.136 |
| MDL Number | MFCD00007984 |
| InChI Key | KWAYEPXDGHYGRW-UHFFFAOYSA-N |
| Synonym | m-nitrobenzamide, benzamide, 3-nitro, benzamide, m-nitro, 3-nitro-benzamide, 3-nitrophenylcarboxamide, acmc-209nmr, 3-nitrobenzamide, dsstox_cid_5732, dsstox_rid_77897, dsstox_gsid_25732 |
| PubChem CID | 12576 |
| IUPAC Name | 3-nitrobenzamide |
| SMILES | C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)N |
Chemical Identifiers
| 645-09-0 | |
| 166.136 | |
| KWAYEPXDGHYGRW-UHFFFAOYSA-N | |
| 12576 | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)N |
| C7H6N2O3 | |
| MFCD00007984 | |
| m-nitrobenzamide, benzamide, 3-nitro, benzamide, m-nitro, 3-nitro-benzamide, 3-nitrophenylcarboxamide, acmc-209nmr, 3-nitrobenzamide, dsstox_cid_5732, dsstox_rid_77897, dsstox_gsid_25732 | |
| 3-nitrobenzamide |
Specifications
| 645-09-0 | |
| Yellow | |
| MFCD00007984 | |
| m-nitrobenzamide, benzamide, 3-nitro, benzamide, m-nitro, 3-nitro-benzamide, 3-nitrophenylcarboxamide, acmc-209nmr, 3-nitrobenzamide, dsstox_cid_5732, dsstox_rid_77897, dsstox_gsid_25732 | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)N | |
| 166.136 | |
| 166.14 | |
| Crystalline Powder |
| 143°C | |
| C7H6N2O3 | |
| 25 g | |
| KWAYEPXDGHYGRW-UHFFFAOYSA-N | |
| 3-nitrobenzamide | |
| 12576 | |
| ≥98.0% (GC) | |
| 3-Nitrobenzamide |
Safety and Handling
RTECSNumber : CV5601250
TSCA : Yes