accessibility menu, dialog, popup

UserName

NH2ONOO

CAS RN 645-09-0

IUPAC Name: 3-nitrobenzamide
Synonyms: M-nitrobenzamide
Molecular Weight (g/mol): 166.14
Molecular Formula: C7H6N2O3
InChi Key: KWAYEPXDGHYGRW-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (5)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)