missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzamide 98.0+%, TCI America™

Catalog No. N032125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N032125G 25 g
N0321500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N032125G Supplier TCI America Supplier No. N032125G

Chemical Identifiers

CAS 645-09-0
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.136
MDL Number MFCD00007984
InChI Key KWAYEPXDGHYGRW-UHFFFAOYSA-N
Synonym m-nitrobenzamide, benzamide, 3-nitro, benzamide, m-nitro, 3-nitro-benzamide, 3-nitrophenylcarboxamide, acmc-209nmr, 3-nitrobenzamide, dsstox_cid_5732, dsstox_rid_77897, dsstox_gsid_25732
PubChem CID 12576
IUPAC Name 3-nitrobenzamide
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)N

Specifications

Melting Point 143°C
Color Yellow
Quantity 25 g
Formula Weight 166.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrobenzamide
RTECSNumber CV5601250
TSCA Yes