missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzamide, 98%

Catalog No. p-172177
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC183440250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC183440250 Supplier Thermo Scientific Chemicals Supplier No. 183440250
Only null left

CAS: 645-09-0 | C7H6N2O3 | 166.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 645-09-0
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.14
MDL Number MFCD00007984
InChI Key KWAYEPXDGHYGRW-UHFFFAOYSA-N
Synonym m-nitrobenzamide, benzamide, 3-nitro, benzamide, m-nitro, 3-nitro-benzamide, 3-nitrophenylcarboxamide, acmc-209nmr, 3-nitrobenzamide, dsstox_cid_5732, dsstox_rid_77897, dsstox_gsid_25732
PubChem CID 12576
IUPAC Name 3-nitrobenzamide
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 140°C to 144°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula O2NC6H4CONH2
Quantity 25 g
Beilstein 09, 381
Solubility Information Solubility in water: slightly soluble
Formula Weight 166.14
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Nitrobenzamide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
EINECSNumber 211-431-0
RTECSNumber CV5601250
TSCA TSCA

RUO – Research Use Only