missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzamide, 98%

Catalog No. AAA1548109
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1548109 10 g
AAA1548118 50 g
AAA1548130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1548109 Supplier Thermo Scientific Chemicals Supplier No. A1548109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 645-09-0 | C7H6N2O3 | 166.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 645-09-0
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.136
MDL Number MFCD00007984
InChI Key KWAYEPXDGHYGRW-UHFFFAOYSA-N
Synonym m-nitrobenzamide, benzamide, 3-nitro, benzamide, m-nitro, 3-nitro-benzamide, 3-nitrophenylcarboxamide, acmc-209nmr, 3-nitrobenzamide, dsstox_cid_5732, dsstox_rid_77897, dsstox_gsid_25732
PubChem CID 12576
IUPAC Name 3-nitrobenzamide
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)N

Specifications

Melting Point 140°C to 143°C
Quantity 10 g
Beilstein 777185
Formula Weight 166.14
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 211-431-0
RTECSNumber CV5601250
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only