Learn More
3-Nitrobenzaldehyde, 99%
CAS: 99-61-6 | C7H5NO3 | 151.121 g/mol
Supplier: Thermo Scientific Chemicals A135940B
Description
3-Nitrobenzaldehyde is used in the synthesis of other organic compounds, ranging from pharmaceuticals to plastic additives and intermediate for the processing of perfume and flavoring compounds and in the preparation of certain aniline dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Nitrobenzaldehyde is used in the synthesis of other organic compounds, ranging from pharmaceuticals to plastic additives and intermediate for the processing of perfume and flavoring compounds and in the preparation of certain aniline dyes.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 3-Nitrobenzaldehyde | |
| 57°C to 59°C | |
| C7H5NO3 | |
| 1000 g | |
| Air Sensitive | |
| m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde | |
| ZETIVVHRRQLWFW-UHFFFAOYSA-N | |
| 3-nitrobenzaldehyde | |
| 7449 | |
| 99% |
| 99-61-6 | |
| 164°C (23 mmHg) | |
| MFCD00007249 | |
| 386795 | |
| 14,6587 | |
| Slightly soluble in water. | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C=O | |
| 151.121 | |
| 151.12 |
Chemical Identifiers
| 99-61-6 | |
| MFCD00007249 | |
| m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde | |
| 3-nitrobenzaldehyde |
| C7H5NO3 | |
| ZETIVVHRRQLWFW-UHFFFAOYSA-N | |
| 7449 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 202-772-6
RTECSNumber : CU7250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only