missing translation for 'onlineSavingsMsg'
Learn More

3-Nitroanisole 99.0+%, TCI America™

Catalog No. N012425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N012425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N012425G Supplier TCI America Supplier No. N012425G
Only null left

Chemical Identifiers

CAS 555-03-3
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007237
InChI Key WGYFINWERLNPHR-UHFFFAOYSA-N
Synonym 3-nitroanisole, m-nitroanisole, benzene, 1-methoxy-3-nitro, m-methoxynitrobenzene, 3-methoxynitrobenzene, anisole, m-nitro, 1-methoxy-3-nitro-benzene, methyl m-nitrophenyl ether, unii-v9920uy61e, ccris 2320
PubChem CID 11140
ChEBI CHEBI:48972
IUPAC Name 1-methoxy-3-nitrobenzene
SMILES COC1=CC=CC(=C1)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 25 g
UN Number 3458
Formula Weight 153.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Nitroanisole
EINECSNumber (3)-0787
RTECSNumber BZ8787000
TSCA Yes
Recommended Storage Refrigerator