Learn More
3-Nitroaniline, 98%
CAS: 99-09-2 | C6H6N2O2 | 138.13 g/mol
Supplier: Thermo Scientific Chemicals A106290E
Description
3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.
Solubility
It is soluble in water 0.1 g/100 ml (20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids, acid chlorides, acid anhydrides, chloroformates, strong oxidizing agents.
Specifications
| 3-Nitroaniline | |
| 112°C to 114°C | |
| 306°C | |
| MFCD00007782 | |
| UN1661 | |
| 14,6582 | |
| It is soluble in water 0.1g/100 ml (20°C). | |
| NC1=CC=CC(=C1)[N+]([O-])=O | |
| 138.13 | |
| 138.13 |
| 99-09-2 | |
| 0.901 | |
| C6H6N2O2 | |
| 2500 g | |
| 636962 | |
| m-nitroaniline, benzenamine, 3-nitro, 3-nitrophenylamine, nitranilin, 3-aminonitrobenzene, fast orange base r, azobase mna, devol orange r, m-nitrophenylamine, 3-nitrobenzenamine | |
| XJCVRTZCHMZPBD-UHFFFAOYSA-N | |
| 3-nitroaniline | |
| 7423 | |
| 98% |
Chemical Identifiers
| 99-09-2 | |
| 138.13 | |
| XJCVRTZCHMZPBD-UHFFFAOYSA-N | |
| 7423 | |
| NC1=CC=CC(=C1)[N+]([O-])=O |
| C6H6N2O2 | |
| MFCD00007782 | |
| m-nitroaniline, benzenamine, 3-nitro, 3-nitrophenylamine, nitranilin, 3-aminonitrobenzene, fast orange base r, azobase mna, devol orange r, m-nitrophenylamine, 3-nitrobenzenamine | |
| 3-nitroaniline |
Safety and Handling
GHS H Statement
H310-H330-H373-H302
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
P260-P262-P264b-P270-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P362-P501c
H301+H311+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROANILINES
EINECSNumber : 202-729-1
RTECSNumber : BY6825000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only