missing translation for 'onlineSavingsMsg'
Learn More

3'-Nitroacetophenone 98.0+%, TCI America™

Catalog No. p-7133623
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N010925G 25 g
N0109100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N010925G Supplier TCI America Supplier No. N010925G
Only null left

Chemical Identifiers

CAS 121-89-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.148
MDL Number MFCD00007259
InChI Key ARKIFHPFTHVKDT-UHFFFAOYSA-N
Synonym 3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon
PubChem CID 8494
IUPAC Name 1-(3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 78°C
Boiling Point 202°C
Quantity 25 g
Formula Weight 165.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Nitroacetophenone
EINECSNumber (3)-1243
RTECSNumber AM9625000
TSCA Yes