missing translation for 'onlineSavingsMsg'
Learn More

3'-Nitroacetophenone, 98+%

Catalog No. AAA1215536
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1215536 500 g
AAA1215530 250 g
AAA121550B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1215536 Supplier Thermo Scientific Chemicals Supplier No. A1215536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 121-89-1 | C8H7NO3 | 165.148 g/mol

3'-Nitroacetophenone is used as an intermediate in pharmaceutical industry for making drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Nitroacetophenone is used as an intermediate in pharmaceutical industry for making drugs.

Solubility
Soluble in water 0.75 g/L @ 20°C.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 121-89-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.148
MDL Number MFCD00007259
InChI Key ARKIFHPFTHVKDT-UHFFFAOYSA-N
Synonym 3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon
PubChem CID 8494
IUPAC Name 1-(3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 76°C to 80°C
Boiling Point 202°C
Odor Slightly sweetish
Quantity 500 g
Beilstein 743002
Solubility Information Soluble in water 0.75g/L @ 20°C.
Formula Weight 165.15
Percent Purity ≥98%
Chemical Name or Material 3'-Nitroacetophenone
EINECSNumber 204-504-3
RTECSNumber AM9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only