missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-L-tyrosine 98.0+%, TCI America™

Catalog No. p-7134022
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09055G 5 g
N090525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09055G Supplier TCI America Supplier No. N09055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 621-44-3
Molecular Formula C9H10N2O5
Molecular Weight (g/mol) 226.19
MDL Number MFCD00007123
InChI Key FBTSQILOGYXGMD-MDOHGIEYNA-N
Synonym 3-nitro-l-tyrosine, 3-nitrotyrosine, h-tyr 3-no2-oh, l-3-nitrotyrosine, tyrosine, 3-nitro, l-tyrosine, 3-nitro, meta-nitro-tyrosine, unii-7coy1ha6hk, tyrosine, 3-nitro-, l, 3-mononitrotyrosine
PubChem CID 65124
ChEBI CHEBI:44454
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3-nitrophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(=C1)[N+]([O-])=O)C(O)=O

Specifications

Melting Point 224°C
Color Yellow
Quantity 5 g
Formula Weight 226.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitro-L-tyrosine