accessibility menu, dialog, popup

UserName

(S)ONH2OHHONOO

CAS RN 621-44-3

IUPAC Name: (2S)-2-amino-3-(4-hydroxy-3-nitrophenyl)propanoic acid
Synonyms: nitrotyrosine
Molecular Weight (g/mol): 226.19
Molecular Formula: C9H10N2O5
InChi Key: FBTSQILOGYXGMD-MDOHGIEYNA-N
SMILES: N[C@@H](CC1=CC=C(O)C(=C1)[N+]([O-])=O)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)