missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-L-tyrosine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1101814

Quantity 25 g

 View more versions of this product

Catalog No. AAA1101814

Only null left

Description

Description

3-Nitro-L-tyrosine is used as an indicator of the formation of peroxynitrite by NO-dependent oxidative damage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitro-L-tyrosine is used as an indicator of the formation of peroxynitrite by NO-dependent oxidative damage.

Solubility
Soluble in water (partly miscible), 1N hydrochloric acid (50 mg/ml), acidic aqueous solution , ethanol, DMSO, DMF and PBS (pH 7.2) (∼2 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

621-44-3
226.19
FBTSQILOGYXGMD-MDOHGIEYNA-N
65124
(2S)-2-amino-3-(4-hydroxy-3-nitrophenyl)propanoic acid
C9H10N2O5
MFCD00007123
3-nitro-l-tyrosine, 3-nitrotyrosine, h-tyr 3-no2-oh, l-3-nitrotyrosine, tyrosine, 3-nitro, l-tyrosine, 3-nitro, meta-nitro-tyrosine, unii-7coy1ha6hk, tyrosine, 3-nitro-, l, 3-mononitrotyrosine
CHEBI:44454
N[C@@H](CC1=CC=C(O)C(=C1)[N+]([O-])=O)C(O)=O

Specifications

∼230°C (decomposition)
621-44-3
MFCD00007123
3-nitro-l-tyrosine, 3-nitrotyrosine, h-tyr 3-no2-oh, l-3-nitrotyrosine, tyrosine, 3-nitro, l-tyrosine, 3-nitro, meta-nitro-tyrosine, unii-7coy1ha6hk, tyrosine, 3-nitro-, l, 3-mononitrotyrosine
FBTSQILOGYXGMD-MDOHGIEYNA-N
(2S)-2-amino-3-(4-hydroxy-3-nitrophenyl)propanoic acid
25 g
CHEBI:44454
98%
3-Nitro-L-tyrosine
C9H10N2O5
2813157
Soluble in water (partly miscible),1N hydrochloric acid (50mg/ml),acidic aqueous solution ,ethanol,DMSO,DMF and PBS (pH 7.2) (∽2mg/ml).
N[C@@H](CC1=CC=C(O)C(=C1)[N+]([O-])=O)C(O)=O
226.19
65124
226.19
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-688-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions