missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-L-tyrosine, 98%, Thermo Scientific Chemicals

Catalog No. AAA1101806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1101806 5 g
AAA1101814 25 g
AAA1101822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1101806 Supplier Thermo Scientific Chemicals Supplier No. A1101806
Only null left

3-Nitro-L-tyrosine is used as an indicator of the formation of peroxynitrite by NO-dependent oxidative damage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitro-L-tyrosine is used as an indicator of the formation of peroxynitrite by NO-dependent oxidative damage.

Solubility
Soluble in water (partly miscible), 1N hydrochloric acid (50 mg/ml), acidic aqueous solution , ethanol, DMSO, DMF and PBS (pH 7.2) (∼2 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 621-44-3
Molecular Formula C9H10N2O5
Molecular Weight (g/mol) 226.19
MDL Number MFCD00007123
InChI Key FBTSQILOGYXGMD-MDOHGIEYNA-N
Synonym 3-nitro-l-tyrosine, 3-nitrotyrosine, h-tyr 3-no2-oh, l-3-nitrotyrosine, tyrosine, 3-nitro, l-tyrosine, 3-nitro, meta-nitro-tyrosine, unii-7coy1ha6hk, tyrosine, 3-nitro-, l, 3-mononitrotyrosine
PubChem CID 65124
ChEBI CHEBI:44454
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3-nitrophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(=C1)[N+]([O-])=O)C(O)=O

Specifications

Melting Point ∼230°C (decomposition)
Beilstein 2813157
Solubility Information Soluble in water (partly miscible),1N hydrochloric acid (50mg/ml),acidic aqueous solution ,ethanol,DMSO,DMF and PBS (pH 7.2) (∽2mg/ml).
Quantity 5 g
Formula Weight 226.19
Percent Purity 98%
Chemical Name or Material 3-Nitro-L-tyrosine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-688-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only