missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-5-(trifluoromethyl)phenol, 98%, Thermo Scientific Chemicals

Catalog No. p-7028708
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2611303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2611303 Supplier Thermo Scientific Chemicals Supplier No. H2611303

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 349-57-5
Molecular Formula C7H4F3NO3
Molecular Weight (g/mol) 207.11
MDL Number MFCD04973779
InChI Key JDIAMHNYAPDMRB-UHFFFAOYSA-N
Synonym 3-nitro-5-trifluoromethyl phenol, 3-hydroxy-5-nitrobenzotrifluoride, 3-trifluoromethyl-5-nitrophenol, phenol, 3-nitro-5-trifluoromethyl, pubchem4155, acmc-1cjq4, 3-nitro-5-trifluoromethyl-phenol, 3-hydroxy-5-trifluoromethyl nitrobenzene
PubChem CID 2756251
IUPAC Name 3-nitro-5-(trifluoromethyl)phenol
SMILES OC1=CC(=CC(=C1)[N+]([O-])=O)C(F)(F)F

Specifications

Melting Point 89°C to 92°C
Quantity 1 g
Formula Weight 207.11
Percent Purity 98%
Chemical Name or Material 3-Nitro-5-(trifluoromethyl)phenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only