missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-5-(trifluoromethyl)benzoic acid, 97%

Catalog No. AAA1831606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1831606 5 g
AAA1831614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1831606 Supplier Thermo Scientific Chemicals Supplier No. A1831606

CAS: 328-80-3 | C8H4F3NO4 | 235.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 328-80-3
Molecular Formula C8H4F3NO4
Molecular Weight (g/mol) 235.118
MDL Number MFCD00024509
InChI Key ODCLHXGXGFBBTA-UHFFFAOYSA-N
Synonym 3-nitro-5-trifluoromethyl benzoic acid, 3-nitro-5-trifluoromethylbenzoic acid, 3-trifluoromethyl-5-nitrobenzoic acid, 3-carboxy-5-nitrobenzotrifluoride, 3-nitro-5 trifluoromethyl benzoic acid, benzoic acid, 3-nitro-5-trifluoromethyl, pubchem4544, acmc-1cr4t, rarechem al bo 1012
PubChem CID 616602
SMILES C1=C(C=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 127°C to 132°C
Quantity 5 g
Beilstein 2284651
Formula Weight 235.12
Percent Purity 97%
Chemical Name or Material 3-Nitro-5-(trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only