missing translation for 'onlineSavingsMsg'
Learn More

3-n-Propylphenol, 98%

Catalog No. AAB2126614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2126614 25 g
AAB2126606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2126614 Supplier Thermo Scientific Chemicals Supplier No. B2126614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 621-27-2 | C9H12O | 136.194 g/mol

3-n-Propylphenol, is used to produce 2,4,5,6-tetrabromo-3-propylphenol. This reaction will need reagent bromine, aluminum powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-n-Propylphenol, is used to produce 2,4,5,6-tetrabromo-3-propylphenol. This reaction will need reagent bromine, aluminum powder.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 621-27-2
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD01632130
InChI Key MPWGZBWDLMDIHO-UHFFFAOYSA-N
Synonym 3-n-propylphenol, m-propylphenol, phenol, 3-propyl, phenol, m-propyl, unii-593r721cic, 1-hydroxy-3-n-propylbenzene, phenol, m-propyl-8ci, phenol, 3-propyl-9ci
PubChem CID 69302
IUPAC Name 3-propylphenol
SMILES CCCC1=CC(=CC=C1)O

Specifications

Boiling Point 229°C to 231°C
Refractive Index 1.525
Quantity 25 g
UN Number UN3145
Solubility Information Soluble in water.
Formula Weight 136.19
Percent Purity 98%
Chemical Name or Material 3-n-Propylphenol
Hazard Category H302+H312-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.
EINECSNumber 210-675-5
RTECSNumber SM8594000
TSCA Yes

RUO – Research Use Only