missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-n-Octylpyrrole 98.0+%, TCI America™
Supplier: TCI America O02415G
Specifications
| 3-n-Octylpyrrole | |
| 111°C | |
| MFCD00191474 | |
| WFHVTZRAIPYMMO-UHFFFAOYSA-N | |
| 3-octyl-1H-pyrrole | |
| 14510078 | |
| ≥98.0% (GC) |
| 118799-18-1 | |
| C12H21N | |
| 5 g | |
| CCCCCCCCC1=CNC=C1 | |
| 179.31 | |
| 179.31 | |
| Liquid |
Chemical Identifiers
| 118799-18-1 | |
| 179.31 | |
| WFHVTZRAIPYMMO-UHFFFAOYSA-N | |
| 3-octyl-1H-pyrrole |
| C12H21N | |
| MFCD00191474 | |
| 14510078 | |
| CCCCCCCCC1=CNC=C1 |
Safety and Handling
TSCA : No