accessibility menu, dialog, popup

UserName

CAS RN 118799-18-1

H3CNH

CAS RN 118799-18-1

IUPAC Name: 3-octyl-1H-pyrrole
Synonyms: 3-octyl-1H-pyrrole
Molecular Weight (g/mol): 179.31
Molecular Formula: C12H21N
InChi Key: WFHVTZRAIPYMMO-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CNC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results