missing translation for 'onlineSavingsMsg'
Learn More

3-n-Octylpyrrole 98.0+%, TCI America™

Catalog No. O02411G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02411G 1 g
O02415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02411G Supplier TCI America Supplier No. O02411G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118799-18-1
Molecular Formula C12H21N
Molecular Weight (g/mol) 179.31
MDL Number MFCD00191474
InChI Key WFHVTZRAIPYMMO-UHFFFAOYSA-N
PubChem CID 14510078
IUPAC Name 3-octyl-1H-pyrrole
SMILES CCCCCCCCC1=CNC=C1

Specifications

Boiling Point 111°C
Quantity 1 g
Formula Weight 179.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-n-Octylpyrrole