missing translation for 'onlineSavingsMsg'
Learn More

3-(N,N-Dimethylpalmitylammonio)propanesulfonate, 98%

Catalog No. AC458500050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458500050 5 g
AC458500250 25 g
AC458501000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC458500050 Supplier Thermo Scientific Chemicals Supplier No. 458500050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2281-11-0 | C21H45NO3S | 391.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2281-11-0
Molecular Formula C21H45NO3S
Molecular Weight (g/mol) 391.66
MDL Number MFCD00036911
InChI Key TUBRCQBRKJXJEA-UHFFFAOYSA-N
Synonym 3-n,n-dimethylpalmitylammonio propanesulfonate, zwittergent 3-16, 3-hexadecyldimethylammonio propane-1-sulfonate, palmityl sulfobetaine, n-hexadecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-hexadecyl dimethyl ammonio propane-1-sulfonate, 3-palmityldimethylammonio propanesulfonate, 3-hexadecyldimethylammonio propanesulfonate, 3-n,n-dimethylhexadecylammonio propanesulfonate, n-hexadecy-n,n-dimethyl-3-ammonio-1-propanesulfonate
PubChem CID 92176
IUPAC Name 3-[hexadecyl(dimethyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

Specifications

Elemental Analysis 3.5 to 3.7% (N, anhydrous) 63.1 to 65.7% (C, anhydrous)
Loss on Drying 2% max.
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information Solubility in water: 100g/L.
Formula Weight 391.65
Percent Purity 98%
Chemical Name or Material 3-(N,N-Dimethylpalmitylammonio)propanesulfonate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 218-918-7

RUO – Research Use Only