missing translation for 'onlineSavingsMsg'
Learn More

3-(N,N-Dimethylmyristylammonio)propanesulfonate, 97%

Catalog No. p-4089814
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427740050 5 g Glass bottle
AC427740250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427740050 Supplier Thermo Scientific Chemicals Supplier No. 427740050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14933-09-6 | C19H41NO3S | 363.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 14933-09-6
Molecular Formula C19H41NO3S
Molecular Weight (g/mol) 363.61
MDL Number MFCD00036910
InChI Key BHATUINFZWUDIX-UHFFFAOYSA-N
Synonym myristyl sulfobetaine, zwittergent 3-14, n-tetradecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dimethyl tetradecyl ammonio propane-1-sulfonate, 3-n,n-dimethylmyristylammonio propanesulfonate, dimethyl 3-sulphonatopropyl tetradecylammonium, zwittergent™ 3-14 detergent, 3-myristyldimethylammonio propanesulfonate, 3-n,n-dimethyltetradecylammonio propanesulfonate
PubChem CID 84705
ChEBI CHEBI:10129
IUPAC Name 3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 246.0°C to 248.0°C
Color Blue to Purple
Elemental Analysis 60.57 to 64.96 % C 10.97 to 11.76 % H 3.72 to 3.99 % N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 363.61
Percent Purity 97%
Physical Form Fine Powder
Chemical Name or Material 3-(N, N-Dimethylmyristylammonio)propanesulfonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
EINECSNumber 239-003-9

RUO – Research Use Only