missing translation for 'onlineSavingsMsg'
Learn More

[3-(N,N-Dimethylamino)propyl]trimethoxysilane 96.0+%, TCI America™

Catalog No. d432825ml
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D432825ML 25 mL
D43285ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D432825ML Supplier TCI America Supplier No. D432825ML
Only null left

Chemical Identifiers

CAS 2530-86-1
Molecular Formula C8H21NO3Si
Molecular Weight (g/mol) 207.35
MDL Number MFCD00053674
InChI Key QIOYHIUHPGORLS-UHFFFAOYSA-N
Synonym N,N-Dimethyl-3-(trimethoxysilyl)propylamine
PubChem CID 75668
IUPAC Name dimethyl[3-(trimethoxysilyl)propyl]amine
SMILES CO[Si](CCCN(C)C)(OC)OC

Specifications

Color Yellow
Boiling Point 42°C
Quantity 25 mL
UN Number 1760
Formula Weight 207.35
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material [3-(N,N-Dimethylamino)propyl]trimethoxysilane