missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[3-(N,N-Dimethylamino)propyl]trimethoxysilane 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 42°C |
| Quantity | 5 mL |
| UN Number | 1760 |
| Formula Weight | 207.35 |
| Percent Purity | ≥96.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | [3-(N,N-Dimethylamino)propyl]trimethoxysilane |
Safety and Handling
| TSCA | Yes |